Stampa…

Hydrocinchonine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (1S)-((2R,4S,5R)-5-Ethylquinuclidin-2-yl)(quinolin-4-yl)methanol, Dihydrocinchonine

H1701-1G H1701-5G
TCIAH1701-1GEA 127 EUR
TCIAH1701-1G TCIAH1701-5G
Hydrocinchonine ≥98.0% (by HPLC, titration analysis)
Hydrocinchonine
Formula: C₁₉H₂₄N₂O
Peso molecolare: 296,41 g/mol
Punto di fusione: 269°C
MDL Number: MFCD00216716
CAS Number: 485-65-4
EINECS: 207-621-8
Merck Index: 14,04783

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +194 to +199 deg (C=0.3, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR