Stampa…

Hexaethylene Glycol Di-p-toluenesulphonate ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl bis(4-methylbenzenesulphonate), Bis-Tos-PEG6, Hexaethylene Glycol Ditosylate

H1681-1G H1681-5G
TCIAH1681-1GEA 52 EUR
TCIAH1681-1G TCIAH1681-5G
Hexaethylene Glycol Di-p-toluenesulphonate ≥95.0% (by HPLC, titration analysis)
Hexaethylene Glycol Di-p-toluenesulphonate
Formula: C₂₆H₃₈O₁₁S₂
MDL Number: MFCD00319275
CAS Number: 42749-27-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 95.0 area%
Purity (Ester Value) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR