Stampa…

1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-(2’-hydroxylethyl)-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide

H1670-5G H1670-25G
TCIAH1670-5GEA 63 EUR
TCIAH1670-5G TCIAH1670-25G
1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulphonyl)imide
Formula: C₈H₁₁F₆N₃O₅S₂
MDL Number: MFCD09038874
CAS Number: 174899-86-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR