Hexabutoxy-mu-oxodititanium(IV) ≥97.0% (W)
Fornitore: TCI EUROPE
|
Attenzione
|
H1668-25G
H1668-500G
TCIAH1668-25GEA
16
EUR
TCIAH1668-25G
TCIAH1668-500G
Hexabutoxy-mu-oxodititanium(IV) ≥97.0% (W)
Hexabutoxy-mu-oxodititanium(IV)
Formula:
C₂₄H₅₄O₇Ti₂ Peso molecolare: 550,42 g/mol |
CAS Number:
7393-46-6 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | White to Yellow to Green clear liquid |
Purity (with ignition residue) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...