Stampa…

4-Hydrazinobenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Phenylhydrazine-4-sulphonic acid, p-Hydrazinobenzenesulphonic acid

H1662-25G H1662-100G H1662-500G
TCIAH1662-25GEA 27 EUR
TCIAH1662-25G TCIAH1662-100G TCIAH1662-500G
4-Hydrazinobenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
4-Hydrazinobenzenesulphonic acid
Formula: C₆H₈N₂O₃S
Peso molecolare: 188,201 g/mol
Punto di fusione: 286°C
MDL Number: MFCD00025097
CAS Number: 98-71-5
EINECS: 202-694-2
Merck Index: 14,04773
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=592cd6b6bbb03e3ef14bfb6da0f769836646239e&vid=592cd6b6bbb03e3ef14bfb6da0f769836646239e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Drying loss max. 2.0 %
Suitability for p-cresol-analysis to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR