Hispidin ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₃H₁₀O₅ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD22199534 CAS Number: 555-55-5 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 60.50 to 66.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...