(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(3R,4S)-3-Hydroxy-4-phenylazetidin-2-one
H1656-250MG
H1656-1G
TCIAH1656-250MGEA
59
EUR
TCIAH1656-250MG
TCIAH1656-1G
(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone ≥98.0% (by HPLC, total nitrogen)
(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone
Formula:
C₉H₉NO₂ Peso molecolare: 163,18 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD07368374 CAS Number: 132127-34-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 186.0 to 190.0 deg-C |
Specific rotation [a]20/D | +174 to +193 deg (C=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...