Stampa…

2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one ≥98.0% (by GC)

Fornitore: TCI

H1653-1G H1653-5G
TCIAH1653-1GEA 64 EUR
TCIAH1653-1G TCIAH1653-5G
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one ≥98.0% (by GC)
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one
Formula: C₁₄H₉NO₃
MDL Number: MFCD02234511
CAS Number: 1218-69-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 204.0 to 208.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR