5-Hydroxy-1H-indazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1H-Indazol-5-ol
H1636-1G
H1636-5G
TCIAH1636-1GEA
34
EUR
TCIAH1636-1G
TCIAH1636-5G
5-Hydroxy-1H-indazole ≥98.0% (by GC)
5-Hydroxy-1H-indazole
Formula:
C₇H₆N₂O Peso molecolare: 134,14 g/mol |
MDL Number:
MFCD01938124 CAS Number: 15579-15-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...