2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol ≥98.0% (by GC)
Fornitore: TCI
H1628-200MG
H1628-50MG
TCIAH1628-200MGEA
207
EUR
TCIAH1628-200MG
TCIAH1628-50MG
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol ≥98.0% (by GC)
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol
Formula:
C₁₄H₁₀O₂S |
MDL Number:
MFCD16619490 CAS Number: 63676-22-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...