10-Hexylphenothiazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
H1608-1G
H1608-5G
TCIAH1608-1GEA
104
EUR
TCIAH1608-1G
TCIAH1608-5G
10-Hexylphenothiazine ≥98.0% (by GC)
10-Hexylphenothiazine
Formula:
C₁₈H₂₁NS Peso molecolare: 283,43 g/mol |
CAS Number:
73025-93-1 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...