Stampa…

6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

H1606-1G H1606-5G
TCIAH1606-1GEA 40 EUR
TCIAH1606-1G TCIAH1606-5G
6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥98.0% (by HPLC, titration analysis)
6-Hydroxy-7-methoxyquinazolin-4(1H)-one
Formula: C₉H₈N₂O₃
MDL Number: MFCD20547768
CAS Number: 179688-52-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR