Stampa…

4-Acetamidobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI

Synonyms: N-[4-(Hydroxymethyl)phenyl]acetamide, 4-Acetamidobenzyl alcohol, N-(4-(Hydroxymethyl)phenyl)acetamide

H1605-1G H1605-5G
TCIAH1605-1GEA 43 EUR
TCIAH1605-1G TCIAH1605-5G
4-Acetamidobenzyl alcohol ≥98.0% (by GC)
4-Acetamidobenzyl alcohol
Formula: C₉H₁₁NO₂
MDL Number: MFCD00016868
CAS Number: 16375-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef1c48edbfae9866ec3cda864370531455e6fac&vid=6ef1c48edbfae9866ec3cda864370531455e6fac&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR