Stampa…

2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, 2-Methyl-6-(trifluoromethyl)-4-pyrimidinol

H1602-1G H1602-5G
TCIAH1602-1GEA 42 EUR
TCIAH1602-1G TCIAH1602-5G
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥98.0% (by GC)
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol
Formula: C₆H₅F₃N₂O
Peso molecolare: 178,1119 g/mol
MDL Number: MFCD03265321
CAS Number: 2836-44-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8f9068f36815b4e405ed1e6c38559f913eb2f62&vid=c8f9068f36815b4e405ed1e6c38559f913eb2f62&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR