Stampa…

4-Hydroxy-1-thiocoumarin ≥98.0% (by GC)

Fornitore: TCI
Attenzione

H1597-1G H1597-5G
TCIAH1597-1GEA 101 EUR
TCIAH1597-1G TCIAH1597-5G
4-Hydroxy-1-thiocoumarin ≥98.0% (by GC)
4-Hydroxy-1-thiocoumarin
Formula: C₉H₆O₂S
MDL Number: MFCD00463527
CAS Number: 16854-67-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 213.0 to 217.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR