Stampa…

3-Amino-5-methylcaproic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: (+/-)-3-Amino-5-methylhexanoic acid, DL-beta-Homoleucine, 3-Amino-5-methylhexanoic acid

H1587-1G H1587-5G
TCIAH1587-1GEA 60 EUR
TCIAH1587-1G TCIAH1587-5G
3-Amino-5-methylcaproic acid ≥98.0% (by titrimetric analysis)
3-Amino-5-methylcaproic acid
Formula: C₇H₁₅NO₂
Peso molecolare: 145,2 g/mol
MDL Number: MFCD01863281
CAS Number: 3653-34-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -1.4 to +1.4 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR