Stampa…
5-Hydroxy-o-toluic acid ≥98.0% (by GC)
Fornitore: TCI
Attenzione
Synonyms: 5-Hydroxy-2-methylbenzoic acid
H1586-1G H1586-5G
TCIAH1586-1GEA 30 EUR
TCIAH1586-1G TCIAH1586-5G
5-Hydroxy-o-toluic acid ≥98.0% (by GC)
5-Hydroxy-o-toluic acid
Formula: C₈H₈O₃
Peso molecolare: 152.15 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09833184
CAS Number: 578-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e8506aae615174356eb1b25221be42a07e46d63&vid=1e8506aae615174356eb1b25221be42a07e46d63&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 184.0 to 188.0 deg-C