2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine
H1574-1G
H1574-5G
TCIAH1574-1GEA
39
EUR
TCIAH1574-1G
TCIAH1574-5G
2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
2-Hydrazino-4-(trifluoromethyl)pyrimidine
Formula:
C₅H₅F₃N₄ Peso molecolare: 178,118 g/mol |
MDL Number:
MFCD00115112 CAS Number: 197305-97-8 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...