Stampa…

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2,3,4-Tetrahydroisoquinolin-6-ol

H1572-1G H1572-5G
TCIAH1572-1GEA 87 EUR
TCIAH1572-1G TCIAH1572-5G
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC)
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Formula: C₉H₁₁NO
Peso molecolare: 149,193 g/mol
MDL Number: MFCD01717047
CAS Number: 14446-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f437b7839b85f379e2829e6632d511b9efcdcdc&vid=6f437b7839b85f379e2829e6632d511b9efcdcdc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR