4-Hydroxy-7-methylcoumarin ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₈O₃ |
MDL Number:
MFCD03412316 CAS Number: 18692-77-8 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...