3',5'-Dimethyl-2'-hydroxyacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Acetyl-4,6-dimethylphenol, 2'-Hydroxy-3',5'-dimethylacetophenone
H1569-1G
H1569-5G
TCIAH1569-1GEA
38
EUR
TCIAH1569-1G
TCIAH1569-5G
3',5'-Dimethyl-2'-hydroxyacetophenone ≥98.0% (by GC)
3',5'-Dimethyl-2'-hydroxyacetophenone
Formula:
C₁₀H₁₂O₂ Peso molecolare: 164,2000 g/mol Punto di ebollizione: 126°C |
MDL Number:
MFCD01631133 CAS Number: 1198-66-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...