Stampa…

4-(Heptyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid

H1566-1G H1566-5G
TCIAH1566-1GEA 42 EUR
TCIAH1566-1G TCIAH1566-5G
4-(Heptyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Heptyloxy)phenylboronic acid
Formula: C₁₃H₂₁BO₃
MDL Number: MFCD04039169
CAS Number: 136370-19-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0586db9bc722e6bb21b278f4c386bb3327bee26a&vid=0586db9bc722e6bb21b278f4c386bb3327bee26a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR