Stampa…

3-Hydroxymethyl-4-methoxybenzeneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Hydroxymethyl)-4-methoxybenzeneboronic acid, 3-(Hydroxymethyl)-4-methoxyphenylboronic acid, (3-(Hydroxymethyl)-4-methoxyphenyl)boronic acid, 3-Hydroxymethyl-4-methoxyphenylboronic acid

H1563-1G H1563-5G
TCIAH1563-1GEA 34 EUR
TCIAH1563-1G TCIAH1563-5G
3-Hydroxymethyl-4-methoxybenzeneboronic acid (contains varying amounts of Anhydride)
3-Hydroxymethyl-4-methoxybenzeneboronic acid
Formula: C₈H₁₁BO₄
Peso molecolare: 181,982 g/mol
MDL Number: MFCD14155851
CAS Number: 908142-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6f2aaf960a3f539a8bc0fac108d062f7f57f8fb&vid=d6f2aaf960a3f539a8bc0fac108d062f7f57f8fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR