Resorcinol monobenzoate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Benzoic acid 3-Hydroxyphenyl ester, 3-Hydroxyphenyl Benzoate, Resorcinol monobenzoate
H1560-25G
H1560-100G
TCIAH1560-25GEA
16
EUR
TCIAH1560-25G
TCIAH1560-100G
Resorcinol monobenzoate ≥95.0% (by GC)
Resorcinol monobenzoate
Formula:
C₁₃H₁₀O₃ Peso molecolare: 214,22 g/mol |
MDL Number:
MFCD00020118 CAS Number: 136-36-7 EINECS: 205-241-7 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 131.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...