4-Hydroxybenzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI
H1547-1G
H1547-250MG
TCIAH1547-1GEA
209
EUR
TCIAH1547-1G
TCIAH1547-250MG
4-Hydroxybenzimidazole ≥98.0% (by GC, titration analysis)
4-Hydroxybenzimidazole
Formula:
C₇H₆N₂O |
MDL Number:
MFCD00963978 CAS Number: 67021-83-4 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...