Stampa…

1-Hexen-5-yne ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

H1541-1G H1541-5G
TCIAH1541-1GEA 155 EUR
TCIAH1541-1G TCIAH1541-5G
1-Hexen-5-yne ≥97.0% (by GC)
1-Hexen-5-yne
Formula: C₆H₈
Peso molecolare: 80,13 g/mol
MDL Number: MFCD00060909
CAS Number: 14548-31-3
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=758cb9d7c598c8f4e8fa88ad880aa97c59d866e2&vid=758cb9d7c598c8f4e8fa88ad880aa97c59d866e2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR