Stampa…
4-Hydroxy-3,5-diiodobenzaldehyde ≥97.0% (by titrimetric analysis)
Fornitore: TCI
Attenzione
Synonyms: 3,5-Diiodo4-hydroxybenzaldehyde, 3,5-Diiodo-4-hydroxybenzaldehyde
H1539-25G H1539-5G
TCIAH1539-25GEA 146 EUR
TCIAH1539-25G TCIAH1539-5G
4-Hydroxy-3,5-diiodobenzaldehyde ≥97.0% (by titrimetric analysis)
4-Hydroxy-3,5-diiodobenzaldehyde
Formula: C₇H₄I₂O₂
Peso molecolare: 373.92 g/mol
MDL Number: MFCD00014670
CAS Number: 1948-40-9
EINECS: 217-754-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b6e35c7dc07bee6359934eacb1d7f117af1948f&vid=5b6e35c7dc07bee6359934eacb1d7f117af1948f&prid=web&clid=SAPDAM
Visualizza i codici
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 202.0 to 206.0 deg-C