Heptylphosphonic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₇H₁₇O₃P |
MDL Number:
MFCD00015840 CAS Number: 4721-16-8 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...