3-Hydroxy-6-methoxyflavone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-Hydroxy-6-methoxy-2-phenyl-4H-chromen-4-one, 3-Hydroxy-6-methoxy-2-phenyl-4-benzopyrone, 6-METHOXYFLAVONOL, 3-Hydroxy-6-methoxy-2-phenylchromen-4-one
H1522-1G
H1522-5G
TCIAH1522-1GEA
48
EUR
TCIAH1522-1G
TCIAH1522-5G
3-Hydroxy-6-methoxyflavone ≥98.0% (by GC)
3-Hydroxy-6-methoxyflavone
Formula:
C₁₆H₁₂O₄ Peso molecolare: 268,27 g/mol |
MDL Number:
MFCD00017639 CAS Number: 93176-00-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 206.0 to 210.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...