Igromicina B ≥80.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
(3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
Formula:
C₂₀H₃₇N₃O₁₃ Punto di fusione: 180°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD06795479 CAS Number: 31282-04-9 EINECS: 250-545-5 UN: 3462 ADR: 6.1,I Merck Index: 14,04852 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) (CAD) | min. 80.0 area% |
Specific rotation [a]20/D | +18.0 to +23.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...