Stampa…

3-Hydroxy-4-iodobenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

H1496-1G H1496-5G
TCIAH1496-1GEA 68 EUR
TCIAH1496-1G TCIAH1496-5G
3-Hydroxy-4-iodobenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-4-iodobenzoic acid
Formula: C₇H₅IO₃
Peso molecolare: 264,02 g/mol
MDL Number: MFCD02068386
CAS Number: 58123-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57d974a512c319e1f0921d740eeb02dc074a27b4&vid=57d974a512c319e1f0921d740eeb02dc074a27b4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 233.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR