Stampa…
2'-Hydroxy-5'-methylpropiophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
H1494-1G H1494-5G
TCIAH1494-1GEA 23 EUR
TCIAH1494-1G TCIAH1494-5G
2'-Hydroxy-5'-methylpropiophenone ≥98.0% (by GC)
2'-Hydroxy-5'-methylpropiophenone
Formula: C₁₀H₁₂O₂
Punto di fusione: 2°C
MDL Number: MFCD01098830
CAS Number: 938-45-4
EINECS: 213-342-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a80de649a3fab6ffdd74e80a1bd844b4e1e2c97&vid=3a80de649a3fab6ffdd74e80a1bd844b4e1e2c97&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %