Stampa…

1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-(2-Hydroxyethyl)-3-methyl-1H-imidazol-3-ium tetrafluoroborate

H1492-5G H1492-25G
TCIAH1492-5GEA 35 EUR
TCIAH1492-5G TCIAH1492-25G
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate
Formula: C₆H₁₁BF₄N₂O
Peso molecolare: 213,97 g/mol
MDL Number: MFCD12761438
CAS Number: 374564-83-7
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bdd4b59c5d391197003ea0c4891944e89803672&vid=0bdd4b59c5d391197003ea0c4891944e89803672&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR