Stampa…

20-Hydroxyecdysone ≥97.0% (by HPLC)

Fornitore: TCI
Attenzione

H1480-100MG H1480-25MG
TCIAH1480-100MGEA 183 EUR
TCIAH1480-100MG TCIAH1480-25MG
20-Hydroxyecdysone ≥97.0% (by HPLC)
20-Hydroxyecdysone
Formula: C₂₇H₄₄O₇
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00036740
CAS Number: 5289-74-7
UN: 3077
ADR: 9,III
Merck Index: 14,03491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2b6adec330f9b99c1c4d2a4926428dcb052d50a&vid=f2b6adec330f9b99c1c4d2a4926428dcb052d50a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +58.0 to +62.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR