(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid
H1477-1G
H1477-5G
TCIAH1477-1GEA
65
EUR
TCIAH1477-1G
TCIAH1477-5G
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Formula:
C₁₀H₁₂O₅ Peso molecolare: 212,2 g/mol |
MDL Number:
MFCD00016834 CAS Number: 4385-56-2 EINECS: 224-494-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...