Stampa…

5-Hydroxynorbornane-2,6-lactone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one

H1475-5G H1475-25G
TCIAH1475-5GEA 29 EUR
TCIAH1475-5G TCIAH1475-25G
5-Hydroxynorbornane-2,6-lactone ≥98.0% (by GC)
5-Hydroxynorbornane-2,6-lactone
Formula: C₈H₁₀O₃
Peso molecolare: 154,17 g/mol
MDL Number: MFCD00988184
CAS Number: 92343-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5c3e8d165b986d300674b35fbfa08c4f899346&vid=be5c3e8d165b986d300674b35fbfa08c4f899346&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR