5-Hydroxypicolinic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-hydroxypyridine-2-carboxylic acid
H1471-1G
H1471-5G
TCIAH1471-1GEA
87
EUR
TCIAH1471-1G
TCIAH1471-5G
5-Hydroxypicolinic acid ≥98.0% (by HPLC, titration analysis)
5-Hydroxypicolinic acid
Formula:
C₆H₅NO₃ Peso molecolare: 139,11 g/mol Punto di fusione: 268°C |
MDL Number:
MFCD00661309 CAS Number: 15069-92-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...