Stampa…

4-Hydroxyphenylretinamide ≥98.0% (by HPLC)

Fornitore: TCI
Pericolo

H1464-10MG H1464-100MG
TCIAH1464-10MGEA 41 EUR
TCIAH1464-10MG TCIAH1464-100MG
4-Hydroxyphenylretinamide ≥98.0% (by HPLC)
4-Hydroxyphenylretinamide
Formula: C₂₆H₃₃NO₂
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00792674
CAS Number: 65646-68-6
Merck Index: 14,03998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a37b75f5d72bd48dc6c5469239813a4bae83726e&vid=a37b75f5d72bd48dc6c5469239813a4bae83726e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 3.40 to 3.80 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR