2-Methyl-6-quinolinol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Hydroxy-2-methylquinoline
H1446-1G
H1446-5G
TCIAH1446-1GEA
86
EUR
TCIAH1446-1G
TCIAH1446-5G
2-Methyl-6-quinolinol ≥98.0% (by GC, titration analysis)
2-Methyl-6-quinolinol
Formula:
C₁₀H₉NO Peso molecolare: 159,19 g/mol |
MDL Number:
MFCD00047600 CAS Number: 613-21-8 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 214.0 to 218.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...