Stampa…

7-Hydroxy-4-(trifluoromethyl)coumarin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Trifluoromethyl)umbelliferone, TFMU

H1422-1G H1422-5G
TCIAH1422-1GEA 60 EUR
TCIAH1422-1G TCIAH1422-5G
7-Hydroxy-4-(trifluoromethyl)coumarin ≥98.0% (by HPLC, titration analysis)
7-Hydroxy-4-(trifluoromethyl)coumarin
Formula: C₁₀H₅F₃O₃
Peso molecolare: 230,14 g/mol
MDL Number: MFCD00037578
CAS Number: 575-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44bca48bdc37bed5c580cfbe397d339ae9daa0ec&vid=44bca48bdc37bed5c580cfbe397d339ae9daa0ec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR