Stampa…

4-Hydroxy-6-methylcoumarin ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

H1421-1G H1421-5G
TCIAH1421-1GEA 40 EUR
TCIAH1421-1G TCIAH1421-5G
4-Hydroxy-6-methylcoumarin ≥98.0% (by GC)
4-Hydroxy-6-methylcoumarin
Formula: C₁₀H₈O₃
Peso molecolare: 176,17 g/mol
Punto di fusione: 261°C
MDL Number: MFCD00085738
CAS Number: 13252-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296656aa9fa30eafb9af506760c612a340783249&vid=296656aa9fa30eafb9af506760c612a340783249&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR