(-)-Corey lactone 98,0 GC_ASSAY_METHOD
Fornitore: TCI
Synonyms:
(-)-6β-Hydroxymethyl-7α-hydroxy-cis-2-oxabicyclo[3.3.0]octan-3-one, (3aR,4S,5R,6aS)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)-Corey lactone diol
Formula:
C₈H₁₂O₄ Peso molecolare: 172.18 g/mol |
MDL Number:
MFCD00075234 CAS Number: 32233-40-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Specific rotation [a]20/D | -39.0 to -43.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...