Stampa…

6-Hydroxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: (6-Hydroxynaphthalen-2-yl)boronic acid, 6-Hydroxynaphthalene-2-boronic acid

H1402-1G H1402-5G
TCIAH1402-1GEA 59 EUR
TCIAH1402-1G TCIAH1402-5G
6-Hydroxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
6-Hydroxy-2-naphthaleneboronic acid
Formula: C₁₀H₉BO₃
Peso molecolare: 187.99 g/mol
MDL Number: MFCD00092986
CAS Number: 173194-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89a05a5efa6e957da4ae4337d9c694ef722cdfc9&vid=89a05a5efa6e957da4ae4337d9c694ef722cdfc9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR