L(+)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Oxfenicine, H-Phg(4-OH)-OH
H1389-5G
H1389-25G
TCIAH1389-5GEA
84
EUR
TCIAH1389-5G
TCIAH1389-25G
L(+)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
L(+)-α-(4-Hydroxyphenyl)glycine
Formula:
C₈H₉NO₃ Peso molecolare: 167,1600 g/mol Punto di fusione: 214°C |
MDL Number:
MFCD00065932 CAS Number: 32462-30-9 EINECS: 251-061-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +155 to +160 deg (C=1、1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...