Stampa…

6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 6-Hydroxy-3-pyridazinecarboxylic Acid Monohydrate, 1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

H1387-1G H1387-5G
TCIAH1387-1GEA 25 EUR
TCIAH1387-1G TCIAH1387-5G
6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥98.0% (by HPLC, titration analysis)
6-Hydroxypyridazine-3-carboxylic acid monohydrate
Formula: C₅H₆N₂O₄
Peso molecolare: 158.11 g/mol
MDL Number: MFCD09064936
CAS Number: 306934-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e6eefbeb098cdb078eb93a9133919a2d2b3ccda&vid=3e6eefbeb098cdb078eb93a9133919a2d2b3ccda&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 10.0 to 14.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR