4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol
H1386-1G
H1386-5G
TCIAH1386-1GEA
179
EUR
TCIAH1386-1G
TCIAH1386-5G
4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol ≥98.0% (by GC)
4-Hydroxy-3-methoxy-alpha-methylbenzyl alcohol
Formula:
C₉H₁₂O₃ Peso molecolare: 168,19 g/mol |
MDL Number:
MFCD00238638 CAS Number: 2480-86-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...