Hydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
H1385-1G
H1385-5G
TCIAH1385-1GEA
65
EUR
TCIAH1385-1G
TCIAH1385-5G
Hydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
Hydroxyterephthalic acid
Formula:
C₈H₆O₅ Punto di fusione: 322°C |
MDL Number:
MFCD09835368 CAS Number: 636-94-2 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...