Stampa…

2-Methylpyridin-3-ol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Hydroxy-2-methylpyridine

H1365-1G H1365-5G
TCIAH1365-1GEA 66 EUR
TCIAH1365-1G TCIAH1365-5G
2-Methylpyridin-3-ol ≥98.0% (by GC, titration analysis)
2-Methylpyridin-3-ol
Formula: C₆H₇NO
Peso molecolare: 109,13 g/mol
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77befa8fefec89a89780b95e6a81b216f0d88267&vid=77befa8fefec89a89780b95e6a81b216f0d88267&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR