Stampa…

4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4'-Methyl-2,2'-bipyridine-4-methanol, (4'-Methyl-[2,2'-bipyridin]-4-yl)methanol, 4-Hydroxymethyl-4'-methyl-2,2'-bipyridine

H1347-1G H1347-200MG
TCIAH1347-1GEA 330 EUR
TCIAH1347-1G TCIAH1347-200MG
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl ≥97.0% (by titrimetric analysis)
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl
Formula: C₁₂H₁₂N₂O
Peso molecolare: 200,24 g/mol
MDL Number: MFCD02916382
CAS Number: 81998-04-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR