2-Hydroxyisonicotinic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
H1336-5G
H1336-25G
TCIAH1336-5GEA
100
EUR
TCIAH1336-5G
TCIAH1336-25G
2-Hydroxyisonicotinic acid ≥98.0% (by GC)
2-Hydroxyisonicotinic acid
Formula:
C₆H₅NO₃ Peso molecolare: 139,11 g/mol Punto di fusione: 328°C |
MDL Number:
MFCD01318566 CAS Number: 22282-72-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...